missing translation for 'onlineSavingsMsg'
Learn More

Di-Sodium Tetraborate Decahydrate, Certified AR for Analysis, Fisher Chemical™

Catalog Number p-7073568
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
1303-96-4
B4Na2O7·10H2O
149193
This item is not returnable. View return policy
For a copy of the TSE/BSE statement please contact Technical Support on Tel: 01509 555888 or email our support team for further assistance
 
1303-96-4
B4Na2O7·10H2O
149193

Di-Sodium Tetraborate Decahydrate, Certified AR for Analysis

TRUSTED_SUSTAINABILITY
Melting Point 75°C
Color White
pH 9
Physical Form Powder/Solid
Quantity 250 g
Formula Weight 381.36g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Sodium tetraborate decahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H319-Causes serious eye irritation.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'