Learn More
Di-Sodium Hydrogen Orthophosphate, for Electrophoresis, Dibasic, Anhydrous, Fisher Chemical™
Brand: Fisher Chemical S/P525/53
Code : 25
Additional Details : CAS Number : 7558-79-4 Weight : 0.50000kg
Packaging | Plastic powder jar |
---|---|
Quantity | 500g |
Chemical Identifiers
7558-79-4 | |
141.957 | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
24203 | |
disodium;hydrogen phosphate |
HNa2O4P | |
3496 | |
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
CHEBI:34683 | |
OP(=O)([O-])[O-].[Na+].[Na+] |
Specifications
7558-79-4 | |
3496 | |
BNIILDVGGAEEIG-UHFFFAOYSA-L | |
disodium;hydrogen phosphate | |
24203 | |
141.96g/mol | |
Plastic powder jar | |
8.7 to 9.3 |
HNa2O4P | |
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt | |
OP(=O)([O-])[O-].[Na+].[Na+] | |
141.957 | |
CHEBI:34683 | |
Powder Solid | |
White | |
500g |