missing translation for 'onlineSavingsMsg'
Learn More

Di-Sodium Hydrogen Orthophosphate, for Electrophoresis, Dibasic, Anhydrous, Fisher Chemical™

Catalog Number p-7073591
Click to view available options
Quantity:
3 kg
Packaging:
Plastic keg
This item is not returnable. View return policy
7558-79-4
HNa2O4P
141.96
MFCD00003496
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
24203
CHEBI:34683
disodium hydrogen phosphate
[Na+].[Na+].OP([O-])([O-])=O
This item is not returnable. View return policy
7558-79-4
HNa2O4P
141.96
MFCD00003496
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
24203
CHEBI:34683
disodium hydrogen phosphate
[Na+].[Na+].OP([O-])([O-])=O

Di-Sodium Hydrogen Orthophosphate, for Electrophoresis, Dibasic, Anhydrous

TRUSTED_SUSTAINABILITY
Color White
pH 8.7 to 9.3
Physical Form Powder/Solid
Quantity 3 kg
Formula Weight 141.96g/mol
Packaging Plastic keg
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'