missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-Sodium Hydrogen Orthophosphate, for Electrophoresis, Dibasic, Anhydrous, Fisher Chemical™
Click to view available options
Quantity:
3 kg
Packaging:
Plastic keg
Description
Di-Sodium Hydrogen Orthophosphate, for Electrophoresis, Dibasic, Anhydrous
Specifications
Color | White |
pH | 8.7 to 9.3 |
Physical Form | Powder/Solid |
Quantity | 3 kg |
Formula Weight | 141.96g/mol |
Packaging | Plastic keg |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'