missing translation for 'onlineSavingsMsg'
Learn More

Di(propylene glycol) methyl ether, 99%, pure, mixture of isomers, Thermo Scientific Chemicals

Catalog Number p-4089916
Click to view available options
Quantity:
1 L
5 L
Packaging:
Glass bottle
Plastic drum
34590-94-8
C7H16O3
148.2
MFCD00059604
34590-94-8
C7H16O3
148.2
MFCD00059604

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -80.0°C
Density 0.9500g/mL
Boiling Point 180.0°C
Flash Point 75°C
Assay Percent Range 98.5% min. sum of isomers (GC)
Packaging Glass bottle
Linear Formula CH3O(CH2)3O(CH2)3OH
Quantity 1 L
Merck Index 15, 3384
Specific Gravity 0.95
Solubility Information Solubility in water: soluble. Other solubilities: miscible with benzene
Viscosity 4 mPa.s (25°C)
Formula Weight 148.2
Percent Purity 99%
Grade Pure
Chemical Name or Material Di(propylene glycol) methyl ether, Mixture of isomers
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'