missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dipotassium Hydrogen Orthophosphate Trihydrate, Certified AR for Analysis, Fisher Chemical™
146.31 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 g
1 kg
Description
Dipotassium Hydrogen Orthophosphate Trihydrate, Certified AR for Analysis, Fisher Chemical, Quantity: 1kg, CAS: 16788-57-1, White, Molecular Formula: H7K2O7P, Molecular Weight: 228.219, Packaging: Plastic powder jar, pH: 8.5 to 9.6
Specifications
Color | White |
pH | 8.5 to 9.6 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 228.23g/mol |
Packaging | Plastic powder jar |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'