missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-Potassium Hydrogen Orthophosphate Trihydrate, Certified AR for Analysis, Fisher Chemical™
Click to view available options
Quantity:
500g
1kg
Description
Di-Potassium Hydrogen Orthophosphate Trihydrate, Certified AR for Analysis, Fisher Chemical, Quantity: 1kg, CAS: 16788-57-1, White, Molecular Formula: H7K2O7P, Molecular Weight: 228.219, Packaging: Plastic powder jar, pH: 8.5 to 9.6
Specifications
Color | White |
pH | 8.5 to 9.6 |
Physical Form | Solid |
Quantity | 1kg |
Formula Weight | 228.23g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Potassium phosphate, dibasic trihydrate