missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Di-p-tolyl sulfone, 98+%

Brand:  Alfa Aesar™ A12176.14

Code : NEW

Additional Details : CAS Number : 599-66-6 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11404823

  • £84.80 / 25g


Estimated Shipment date
from Supplier 01-02-2018
Add to basket

Specifications

Chemical Identifiers

Di-p-tolyl sulfone
C14H14O2S
MFCD00041332
599-66-6
246.32
Bis(4-methylphenyl) sulfone

Specifications

Di-p-tolyl sulfone
25g
≥98%
246.32
Bis(4-methylphenyl) sulfone
156° to 161°C
599-66-6
C14H14O2S
MFCD00041332
Videos
SDS