missing translation for 'onlineSavingsMsg'
Learn More

Di-p-tolyl ether, 99%, Thermo Scientific Chemicals

Catalog Number 15441328
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1579-40-4
C14H14O
198.265
MFCD00025979
YWYHGNUFMPSTTR-UHFFFAOYSA-N
p-tolyl ether, di-p-tolyl ether, 4-tolyl ether, 4,4'-oxybis methylbenzene, bis 4-methylphenyl ether, 4,4'-dimethyldiphenyl ether, benzene, 1,1'-oxybis 4-methyl, p-p-tolyloxy toluene, 1,1'-oxybis 4-methylbenzene
74098
1-methyl-4-(4-methylphenoxy)benzene
CC1=CC=C(C=C1)OC2=CC=C(C=C2)C
This item is not returnable. View return policy
1579-40-4
C14H14O
198.265
MFCD00025979
YWYHGNUFMPSTTR-UHFFFAOYSA-N
p-tolyl ether, di-p-tolyl ether, 4-tolyl ether, 4,4'-oxybis methylbenzene, bis 4-methylphenyl ether, 4,4'-dimethyldiphenyl ether, benzene, 1,1'-oxybis 4-methyl, p-p-tolyloxy toluene, 1,1'-oxybis 4-methylbenzene
74098
1-methyl-4-(4-methylphenoxy)benzene
CC1=CC=C(C=C1)OC2=CC=C(C=C2)C

Di-p-tolyl ether is used to form conformationally chiral molecules in the solid state, while the chirality of 1,3-dimethyl-2-phenoxybenzene arises from the formation of supramolecular helices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Di-p-tolyl ether is used to form conformationally chiral molecules in the solid state, while the chirality of 1,3-dimethyl-2-phenoxybenzene arises from the formation of supramolecular helices.

Solubility
Soluble in acetone, benzene, ether.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Melting Point 47°C to 49°C
Color Yellow
Boiling Point 285°C
Flash Point >113°C (235°F)
Quantity 5 g
UN Number UN3077
Solubility Information Soluble in acetone,benzene,ether.
Formula Weight 198.26
Percent Purity 99%
Physical Form Solid
Chemical Name or Material Di-p-tolyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'