missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-p-tolyl ether, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
Di-p-tolyl ether is used to form conformationally chiral molecules in the solid state, while the chirality of 1,3-dimethyl-2-phenoxybenzene arises from the formation of supramolecular helices.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Di-p-tolyl ether is used to form conformationally chiral molecules in the solid state, while the chirality of 1,3-dimethyl-2-phenoxybenzene arises from the formation of supramolecular helices.
Solubility
Soluble in acetone, benzene, ether.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Di-p-tolyl ether is used to form conformationally chiral molecules in the solid state, while the chirality of 1,3-dimethyl-2-phenoxybenzene arises from the formation of supramolecular helices.
Solubility
Soluble in acetone, benzene, ether.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Melting Point | 47°C to 49°C |
Color | Yellow |
Boiling Point | 285°C |
Flash Point | >113°C (235°F) |
Quantity | 25 g |
UN Number | UN3077 |
Solubility Information | Soluble in acetone,benzene,ether. |
Formula Weight | 198.26 |
Percent Purity | 99% |
Physical Form | Solid |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'