missing translation for 'onlineSavingsMsg'
Learn More

(-)-Di-p-toluoyl-L-tartaric acid, 97%, Thermo Scientific Chemicals

Catalog Number 10580741
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
32634-66-5
C20H18O8
386.36
MFCD00064440
CMIBUZBMZCBCAT-IYBDPMFKSA-N
2r,3r-2,3-bis 4-methylbenzoyl oxy succinic acid, di-p-toluoyl-l-tartaric acid, l-dtta, --di-p-toluoyl-l-tartaric acid, --di-1,4-o-toluoyl-l-tartaric acid, --o,o'-di-p-toluoyl-l-tartaric acid, l---di-p-toluoyltartaric acid, 2r,3r-2,3-bis 4-methylbenzoyloxy butanedioic acid, 2r,3r-2,3-bis 4-methylbenzoyl oxy butanedioic acid, butanedioicacid, 2,3-bis 4-methylbenzoyl oxy-, 2r,3r
12297760
(2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;hydrate
CC1=CC=C(C=C1)C(=O)O[C@@H]([C@@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O
This item is not returnable. View return policy
32634-66-5
C20H18O8
386.36
MFCD00064440
CMIBUZBMZCBCAT-IYBDPMFKSA-N
2r,3r-2,3-bis 4-methylbenzoyl oxy succinic acid, di-p-toluoyl-l-tartaric acid, l-dtta, --di-p-toluoyl-l-tartaric acid, --di-1,4-o-toluoyl-l-tartaric acid, --o,o'-di-p-toluoyl-l-tartaric acid, l---di-p-toluoyltartaric acid, 2r,3r-2,3-bis 4-methylbenzoyloxy butanedioic acid, 2r,3r-2,3-bis 4-methylbenzoyl oxy butanedioic acid, butanedioicacid, 2,3-bis 4-methylbenzoyl oxy-, 2r,3r
12297760
(2R,3R)-2,3-bis[(4-methylbenzoyl)oxy]butanedioic acid;hydrate
CC1=CC=C(C=C1)C(=O)O[C@@H]([C@@H](OC(=O)C1=CC=C(C)C=C1)C(O)=O)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point 164°C to 170°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula [CH3C6H4CO2CH(CO2H)-]2
Quantity 10 g
Specific Rotation Condition −142° (20°C c=10,C2H5OH)
Specific Rotation -142
Solubility Information Solubility in water: soluble. Other solubilities: soluble in acetone and ethanol
Formula Weight 386.36
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (-)-Di-p-toluoyl-L-tartaric acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'