missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Di-n-pentyl succinate, 95%

Brand:  Alfa Aesar™ H64644.14

Code : NEW

Additional Details : CAS Number : 645-69-2 Weight : 0.02500kg Transport : UN number : 3082 Chem class : 1 Pack group : III

Quantity 25g

 View more versions of this product

Product Code. 15441929

  • £55.54 / 25g


Estimated Shipment date
from Supplier 21-02-2018
Add to basket

Specifications

Chemical Identifiers

Di-n-pentyl succinate
258.36
UN3082
Succinic acid di-n-pentyl ester; Di-n-pentyl ethane-1,2-dicarboxylate
645-69-2
MFCD00086866
1790389

Specifications

Di-n-pentyl succinate
129°C (2mmHg) (lit.)
25g
1.4435
95%
MFCD00086866
1790389
0.962g/mL
−9°C (lit.)
645-69-2
Liquid
258.36
UN3082
Succinic acid di-n-pentyl ester; Di-n-pentyl ethane-1,2-dicarboxylate
Videos
SDS