missing translation for 'onlineSavingsMsg'
Learn More

Di-n-nonyl sulfide, 98%, Thermo Scientific™

Catalog Number p-7045421
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
929-98-6
C18H38S
286.56
MFCD00027342
KMKSVAGOBVUFRO-UHFFFAOYSA-N
dinonyl sulfide, di-n-nonyl sulfide, nonyl sulfide, 1-nonylsulfanyl nonane, nonylthiononane, dinonylsulfide, n-nonylsulfide, dinonyl sulphide, n-nonyl sulfide, octylmethylsulfide
525006
1-nonylsulfanylnonane
CCCCCCCCCSCCCCCCCCC
This item is not returnable. View return policy
929-98-6
C18H38S
286.56
MFCD00027342
KMKSVAGOBVUFRO-UHFFFAOYSA-N
dinonyl sulfide, di-n-nonyl sulfide, nonyl sulfide, 1-nonylsulfanyl nonane, nonylthiononane, dinonylsulfide, n-nonylsulfide, dinonyl sulphide, n-nonyl sulfide, octylmethylsulfide
525006
1-nonylsulfanylnonane
CCCCCCCCCSCCCCCCCCC

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 15°C
Density 0.855
Boiling Point 165°C to 167°C (3 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Linear Formula CH3(CH2)8S(CH2)8CH3
Refractive Index 1.464
Quantity 5 g
Beilstein 1755874
Formula Weight 286.56
Percent Purity 98%
Chemical Name or Material Di-n-nonyl sulfide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'