missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butylamine, for HPLC, Thermo Scientific Chemicals

Catalog Number p-3586424
Click to view available options
Quantity:
10 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
111-92-2
C8H19N
129.24
MFCD00009429
JQVDAXLFBXTEQA-UHFFFAOYSA-N
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
8148
N-butylbutan-1-amine
CCCCNCCCC
This item is not returnable. View return policy
111-92-2
C8H19N
129.24
MFCD00009429
JQVDAXLFBXTEQA-UHFFFAOYSA-N
dibutylamine, di-n-butylamine, 1-butanamine, n-butyl, n-butyl-1-butanamine, dibutilamina, n-dibutylamine, di-n-butyl amine, dibutyl amine, di-normal-butylamine, dibutyl-amine
8148
N-butylbutan-1-amine
CCCCNCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -62.0°C
Color White
Density 0.7600g/mL
Boiling Point 159.0°C
Flash Point 39°C
Packaging Glass bottle
Linear Formula [CH3(CH2)3]2NH
Refractive Index 1.4160 to 1.4180
Quantity 10 mL
Residue after Evaporation 0.0005% max.
Merck Index 15, 3037
Specific Gravity 0.76
Solubility Information Solubility in water: 4.05g/L (25°C). Other solubilities: soluble in alcohol, ether, benzene, acetone, miscible with hydrocarbons
Transmittance 99 (c=0.01M water, 254nm) min.
Viscosity 0.9 mPa.s (20°C)
Formula Weight 129.24
Percent Purity ≥99% (GC)
Grade HPLC
Physical Form Crystalline Free Flowing Powder
Chemical Name or Material Di-n-butylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'