missing translation for 'onlineSavingsMsg'
Learn More

Di-^m-chlorodichlorobis(ethylene)diplatinum(II), 97%, Thermo Scientific Chemicals

Catalog Number p-7050692
Click to view available options
Quantity:
250 mg
This item is not returnable. View return policy
12073-36-8
C4H8Cl4Pt2
588.08
MFCD00058713
XBLBDYCZFZESPN-UHFFFAOYSA-J
di-mu-chloro-dichlorobis ethylene diplatinum ii, zeise's dimer, ptcl2 c2h4 2, di-chloro-dichlorobis ethylene diplatinum ii, di-micro-chlorodichlorobis ethylene diplatinum ii
11124780
[Cl-].[Cl-].[Cl-].[Cl-].[Pt++].[Pt++].C=C.C=C
This item is not returnable. View return policy
12073-36-8
C4H8Cl4Pt2
588.08
MFCD00058713
XBLBDYCZFZESPN-UHFFFAOYSA-J
di-mu-chloro-dichlorobis ethylene diplatinum ii, zeise's dimer, ptcl2 c2h4 2, di-chloro-dichlorobis ethylene diplatinum ii, di-micro-chlorodichlorobis ethylene diplatinum ii
11124780
[Cl-].[Cl-].[Cl-].[Cl-].[Pt++].[Pt++].C=C.C=C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
IUPAC Name dichloroplatinum;ethene
Formula Weight 588.1
Percent Purity 97%
Physical Form Crystalline
Chemical Name or Material Di-mu-chlorodichlorobis(ethylene)diplatinum(II)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'