missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Di-Iso-PropylnapHthalene, Extra Pure, Mixed Isomers, SLR, Fisher Chemical
Click to view available options
Quantity:
2.5 L
Packaging:
Amber glass bottle
Specifications
CAS | 38640-62-9 |
Melting Point | -47°C |
Color | Colorless |
Boiling Point | 290°C |
Packaging | Amber glass bottle |
Molecular Formula | C16H20 |
MDL Number | 66401 |
Quantity | 2.5 L |
Synonym | 1,5-diisopropylnaphthalene, unii-b091e29zir, naphthalene, 1,5-bis 1-methylethyl, 1,5-di propan-2-yl naphthalene, naphthalene, 1,5-diisopropyl, 1,5-bis isopropyl naphthalene, naphthalene,1,5-bis 1-methylethyl, diisopropylnaphthalene mixture of isomers, unii-v970u16pqr component |
InChI Key | GFZWCYSEPXDDRI-UHFFFAOYSA-N |
Show More |
Safety and Handling
Product Identifier
- Di-iso-propylnaphthalene
- Danger
- Aspiration toxicity Category 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H304-May be fatal if swallowed and enters airways.
- H410-Very toxic to aquatic life with long lasting effects.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P273-Avoid release to the environment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'