missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Di-[3-((S)-2,2'-dihydroxy-1,1'-binaphthylmethyl)]ether, lanthanum(III) salt, tetrahydrofuran adduct, SCT-(S)-BINOL, 95%

Asymmetric Michael reaction

Brand:  Alfa Aesar™ 044611.77

Code : NEW

Additional Details : CAS Number : 293293-33-1 Weight : 0.00010kg

Quantity 0.1g

 View more versions of this product

Product Code. 11475400

  • £198.75 / 0.10g


Estimated Shipment date
from Supplier 05-06-2018
Add to basket

Specifications

Chemical Identifiers

Di-[3- ( (S)-2,2'-dihydroxy-1,1'-binaphthylmethyl)]ether, lanthanum (III) salt, tetrahydrofuran adduct
SCT-(S)-BINOL
822.69
293293-33-1
C42H27LaO5·C4H8O
MFCD03098650

Specifications

Di-[3- ( (S)-2,2'-dihydroxy-1,1'-binaphthylmethyl)]ether, lanthanum (III) salt, tetrahydrofuran adduct
0.1g
293293-33-1
95%
822.69
Pale brown
SCT-(S)-BINOL
Powder
C42H27LaO5·C4H8O
MFCD03098650
Videos
SDS