missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Di(2-pyridyl) ketone thiosemicarbazone, 97%

Brand:  Alfa Aesar™ H65728.03

Code : NEW

Additional Details : CAS Number : 6839-91-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15402619

  • £58.30 / 1g


Estimated Shipment date
from Supplier 04-05-2018
Add to basket

Specifications

Chemical Identifiers

Di(2-pyridyl) ketone thiosemicarbazone
6839-91-4
257.07
MFCD26097263

Specifications

Di(2-pyridyl) ketone thiosemicarbazone
1g
Solid
257.07
Green-yellow
6839-91-4
97%
MFCD26097263
Videos
SDS