Learn More
DHTKD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558180
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EIKSSYYAKL NDHLNNMAHY RPPALNLQAH WQGLAQPEAQ ITTWSTGVPL DLLRFVGMKS VEVPRELQMH SHLLKTHVQS RMEKMMDG Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 74%.
The 2-oxoglutarate dehydrogenase complex catalyzes the overall conversion of 2-oxoglutarate to succinyl-CoA and CO(2). It contains multiple copies of three enzymatic components: 2-oxoglutarate dehydrogenase (E1), dihydrolipoamide succinyltransferase (E2) and lipoamide dehydrogenase (E3).Specifications
DHTKD1 | |
Polyclonal | |
Unconjugated | |
DHTKD1 | |
AMOXAD; C330018I04Rik; CMT2Q; dehydrogenase E1 and transketolase domain containing 1; dehydrogenase E1 and transketolase domain-containing protein 1; Dhtkd1; DKFZp762M115; KIAA1630; MGC3090; Probable 2-oxoglutarate dehydrogenase E1 component DHKTD1, mitochondrial | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55526 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96HY7 | |
DHTKD1 | |
Recombinant protein corresponding to Human DHTKD1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only