GE Healthcare Dharmacon siSTABLE Cyclophilin B Control siRNA - Mouse


Overview
Brand: GE Healthcare

Manufacturer Part Number: D-001710-03-05

SISTABLE CYCLO B SIR(MO) 5NMOL

UNSPSC: 41106402

Code: P1

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11517230

Quantity Price
1 £ 150.22 / 5nmol
Estimated Shipment date
from Supplier 27-03-2017
Add to basket


Description and Specification

Specification

Storage Requirements –20°C
Quantity 5nmol
For Use With (Application) Suitable for use in experimental settings with high nuclease activity, including in vivo studies
Includes In vivo studies