GE Healthcare Dharmacon miRIDIAN™ microRNA Mimic Negative Control #2


Overview
Brand: GE Healthcare

Manufacturer Part Number: IN-002000-01-20

UNSPSC: 41106402

Code: P1

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11511731

Quantity Price
1 £ 684.78 / Each
Estimated Shipment date
from Supplier 06-06-2017
Add to basket


Description and Specification

Specification

Storage Requirements –20°C
Quantity 20nmol
For Use With (Application) Confirmed to have minimal sequence identity with miRNAs in human, mouse, and rat