Learn More
Tocris Bioscience™ DH 97
MT2 receptor antagonist
Brand:  Tocris Bioscience™ 121850
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13428913
  • £568.89 / 50mg
Estimated Shipment date
from Supplier 06-04-2020
Description
Description
A relatively potent MT2 melatonin receptor antagonist (pKi value = 8.03), displaying 89- and 229-fold selectivity over MT1 and GPR50 (melatonin-related orphan receptor) respectively.
Specifications
Specifications
DH 97
DH 97
334.46