Learn More
Tocris Bioscience™ DH 97
MT2 receptor antagonist
POA
Specifications
Product Type DH 97
Molecular Weight (g/mol) 334.46
Inhibitors DH 97
View More Specs
Description
Description
A relatively potent MT2 melatonin receptor antagonist (pKi value = 8.03), displaying 89- and 229-fold selectivity over MT1 and GPR50 (melatonin-related orphan receptor) respectively.
Specifications
Specifications
DH 97
334.46
DH 97