Learn More
DFMO, Tocris Bioscience™
Ornithine decarboxylase inhibitor
Brand:  Tocris (Bio-Techne) 276150
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13450544
  • £286.65 / 50mg
Estimated Shipment date
from Supplier 21-11-2019
Description
Description
Irreversible inhibitor of ornithine decarboxylase (ODC) that inhibits polyamine biosynthesis. Displays antiapoptotic, antiangiogenic and antiparasitic activity. Induces re-expression of aberrantly silenced tumor suppressor genes when used in combination with oligoamine analogs.
Specifications
Chemical Identifiers
68278-23-9
218.629
eflornithine hydrochloride, eflornithine hcl, dfmo hcl, dl-alpha-difluoromethylornithine hydrochloride, dfmo, vaniqa, ccris 5805, 2,5-diamino-2-difluoromethyl pentanoic acid hydrochloride, 2-difluoromethyl-dl-ornithine monohydrochloride
2,5-diamino-2-(difluoromethyl)pentanoic acid;hydrochloride
C6H13ClF2N2O2
VKDGNNYJFSHYKD-UHFFFAOYSA-N
57004
C(CC(C(F)F)(C(=O)O)N)CN.Cl
Specifications
50mg
68278-23-9
eflornithine hydrochloride, eflornithine hcl, dfmo hcl, dl-alpha-difluoromethylornithine hydrochloride, dfmo, vaniqa, ccris 5805, 2,5-diamino-2-difluoromethyl pentanoic acid hydrochloride, 2-difluoromethyl-dl-ornithine monohydrochloride
C(CC(C(F)F)(C(=O)O)N)CN.Cl
218.629
218.63
DFMO
C6H13ClF2N2O2
VKDGNNYJFSHYKD-UHFFFAOYSA-N
2,5-diamino-2-(difluoromethyl)pentanoic acid;hydrochloride
57004
>98%