Learn More
DFB, Tocris Bioscience™
Allosteric potentiator at mGlu5
Brand:  Tocris (Bio-Techne) 162510
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13459823
  • £81.00 / 10mg
Estimated Shipment
04-03-2019
Add to basket
Description
Description
Novel allosteric potentiator of the metabotropic glutamate receptor mGlu5. Devoid of agonist activity itself, but potentiates (3 - 6-fold) the action of agonists at mGlu5 without any effect at other mGlu subtypes (EC50 for potentiation = 2 - 5.3 μM).
Specifications
Chemical Identifiers
15332-10-2
244.245
3,3'-difluorobenzaldazine, 1,2-bis 3-fluorobenzylidene hydrazine, dfb, d0f9qy, bis 3-fluorobenzylidene hydrazine, dfb hplc , solid, z,z-bis 3-fluorophenyl methylidene hydrazine, 3,3 inverted exclamation marka-difluorobenzaldazine, 3-fluorophenyl methylene hydrazone-3-fluorobenzaldehyde, benzaldehyde,3-fluoro-, 2-3-fluorophenyl methylene hydrazone
(E)-1-(3-fluorophenyl)-N-[(E)-(3-fluorophenyl)methylideneamino]methanimine
C14H10F2N2
YYMCVDNIIFNDJK-BEQMOXJMSA-N
6604893
C1=CC(=CC(=C1)F)C=NN=CC2=CC(=CC=C2)F
Specifications
DFB
15332-10-2
3,3'-difluorobenzaldazine, 1,2-bis 3-fluorobenzylidene hydrazine, dfb, d0f9qy, bis 3-fluorobenzylidene hydrazine, dfb hplc , solid, z,z-bis 3-fluorophenyl methylidene hydrazine, 3,3 inverted exclamation marka-difluorobenzaldazine, 3-fluorophenyl methylene hydrazone-3-fluorobenzaldehyde, benzaldehyde,3-fluoro-, 2-3-fluorophenyl methylene hydrazone
C1=CC(=CC(=C1)F)C=NN=CC2=CC(=CC=C2)F
244.245
244.24
10mg
C14H10F2N2
YYMCVDNIIFNDJK-BEQMOXJMSA-N
(E)-1-(3-fluorophenyl)-N-[(E)-(3-fluorophenyl)methylideneamino]methanimine
6604893
>99%
Videos