missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dextran sulfate sodium salt, Reagent grade, MP Biomedicals™
Click to view available options
Quantity:
10 g
100 g
Specifications
Color | White |
Physical Form | Powder |
Quantity | 10 g |
IUPAC Name | ethyl 4-aminobenzoate |
Formula Weight | 8000 (Average) |
Grade | Reagent |
Chemical Name or Material | Dextran sulfate sodium salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'