Learn More
Dexamethasone, Tocris Bioscience™
Anti-inflammatory glucocorticoid
Brand:  Tocris (Bio-Techne) 1126100
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 100mg
Product Code. 13498603
  • £92.61 / 100mg
Estimated Shipment date
from Supplier 26-02-2020
Description
Description
Glucocorticoid; anti-inflammatory. Reduces levels of activated NF-κB in immature dendritic cells (DCs) and inhibits differentiation into mature DCs. Induces differentiation of human mesenchymal stem cells (MSCs). Also induces autophagy in acute lymphoblastic leukemia (ALL) cell lines.
Specifications
Specifications
100mg
50-02-2
dexamethasone, decadron, dexamethazone, maxidex, decaspray, desametasone, hexadecadrol, dexacort, dexasone, hexadrol
CC1CC2C3CCC4=CC(=O)C=CC4(C3(C(CC2(C1(C(=O)CO)O)C)O)F)C
392.467
CHEBI:41879
>98%
Dexamethasone
C22H29FO5
UREBDLICKHMUKA-CXSFZGCWSA-N
(8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
5743
392.47