missing translation for 'onlineSavingsMsg'
Learn More

Deuterium oxide, 99.8% (Isotopic), Thermo Scientific Chemicals

Catalog Number p-4408389
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7789-20-0
H2O
20.03
MFCD00044636
XLYOFNOQVPJJNP-ZSJDYOACSA-N
deuterium oxide, water-d2, heavy water, deuterated water, dideuterium oxide, heavy water-d2, heavy water d2o, water sup 2-h2, deuterium oxide usan, water, heavy d2-o
24602
CHEBI:41981
[(²H)oxy](²H)
[2H]O[2H]
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7789-20-0
H2O
20.03
MFCD00044636
XLYOFNOQVPJJNP-ZSJDYOACSA-N
deuterium oxide, water-d2, heavy water, deuterated water, dideuterium oxide, heavy water-d2, heavy water d2o, water sup 2-h2, deuterium oxide usan, water, heavy d2-o
24602
CHEBI:41981
[(²H)oxy](²H)
[2H]O[2H]

Deuterium oxide is used in nuclear magnetic resonance spectroscopy (NMR). It is also useful in the identification of labile hydrogens. As a source of deuterium, it is utilized for preparing specifically labeled isotopologs of organic compounds. It is often used as a substitute for water in the analysis of proteins in solution by using fourier transform infrared spectroscopy (FTIR). It finds application in certain types of nuclear reactors and in tritium production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless
Density 1.105g/cm3 at 20°C
Odor Odorless
Refractive Index 1.338
Sensitivity Moisture
Vapor Pressure 6 hPa at 25°C
Quantity 50 g
Melting Point 3.82°C
Boiling Point 101.4°C
Assay Percent Range 99.8% (Isotopic)
Solubility Information Miscible with water.
Formula Weight 20.03
Chemical Name or Material Deuterium oxide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'