missing translation for 'onlineSavingsMsg'
Learn More

Deuterium chloride, 20% w/w in D{2}O, 99.96% (Isotopic), Thermo Scientific Chemicals

Catalog Number p-7052022
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7698-05-7
ClH
36.46
MFCD00044642
VEXZGXHMUGYJMC-FTGQXOHASA-N
deuterium chloride, hydrochloric acid-d, deuterium chloride, 20 wt% in deuterium oxide, chlorohydrogen, hydrochloric acid d, chloro 2h, deuterium chloride, 99 atom % d, deuterium chloride, 1 m in deuterium oxide, deuterium chloride, 0.1 m in deuterium oxide, deuterium chloride, 38 wt% in deuterium oxide
522634
Cl
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7698-05-7
ClH
36.46
MFCD00044642
VEXZGXHMUGYJMC-FTGQXOHASA-N
deuterium chloride, hydrochloric acid-d, deuterium chloride, 20 wt% in deuterium oxide, chlorohydrogen, hydrochloric acid d, chloro 2h, deuterium chloride, 99 atom % d, deuterium chloride, 1 m in deuterium oxide, deuterium chloride, 0.1 m in deuterium oxide, deuterium chloride, 38 wt% in deuterium oxide
522634
Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless to Yellow
Density 1.20 g/mL
Odor Acidic
Packaging Clear Glass Ampule
Refractive Index 1.373
Sensitivity Light and Moisture
Quantity 5 g
Boiling Point 108°C
UN Number UN1789
Solubility Information Fully miscible in water.
Formula Weight 37.47
Percent Purity ≥99.96%
Concentration or Composition (by Analyte or Components) 20% w/w in D2O
Chemical Name or Material Deuterium chloride, Isotopic
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'