missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dess-Martin periodinane, Thermo Scientific Chemicals
18.80 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Dess Martin periodinane is used to oxidize primary alcohols to aldehydes and secondary alcohols to ketones. It is also used as an oxidant for complexes with multifunctional alcohols. It is actively involved in the oxidation of N-protected-amino alcohols without epimerization and allylic alcohols.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dess Martin periodinane is used to oxidize primary alcohols to aldehydes and secondary alcohols to ketones. It is also used as an oxidant for complexes with multifunctional alcohols. It is actively involved in the oxidation of N-protected-amino alcohols without epimerization and allylic alcohols.
Solubility
Soluble in chloroform, acetone, acetonitrile and methylene chloride. Slightly soluble in ether and hexane.
Notes
Store in cool place. Air and moisture sensitive. Incompatible with strong reducing agents, combustible material and water.
Dess Martin periodinane is used to oxidize primary alcohols to aldehydes and secondary alcohols to ketones. It is also used as an oxidant for complexes with multifunctional alcohols. It is actively involved in the oxidation of N-protected-amino alcohols without epimerization and allylic alcohols.
Solubility
Soluble in chloroform, acetone, acetonitrile and methylene chloride. Slightly soluble in ether and hexane.
Notes
Store in cool place. Air and moisture sensitive. Incompatible with strong reducing agents, combustible material and water.
Specifications
Melting Point | 130°C to 132°C (decomposition) |
Quantity | 1 g |
UN Number | UN1479 |
Beilstein | 4548207 |
Sensitivity | Air, light, and moisture sensitive |
Merck Index | 14,2933 |
Synonym | dess-martin periodinane, triacetoxyperiodinane, 1,1,1-triacetoxy-1,1-dihydro-1,2-benziodoxol-3 1h-one, dess-martin reagent, dess-martinperiodinane, dess-martin, dess martin periodinane, dess martin, dess-martin periodane |
Solubility Information | Soluble in chloroform,acetone,acetonitrile and methylene chloride. Slightly soluble in ether and hexane. |
IUPAC Name | (1,1-diacetyloxy-3-oxo-1$l^{5},2-benziodoxol-1-yl) acetate |
Formula Weight | 424.15 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'