missing translation for 'onlineSavingsMsg'
Learn More

Desmoglein-3 Mouse, Clone: 5G11, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 1830-MSM3-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15939907

  • £395.00 / 100µg
Estimated Shipment: 16-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Desmoglein-3
Cell surface
5G11
Unconjugated
NP_001935
Mouse
130kDa
Protein A/G Purified
RUO
1830
IgG1 κ
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
DSG3
Recombinant human DSG3 extracellular protein fragment
A431 cells. Lung squamous cell carcinoma.
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers