missing translation for 'onlineSavingsMsg'
Learn More

Desmin, eFluor 660, clone: DE-U-10, eBioscience™

Mouse Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-9747-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15530117

  • £172.75 / 100µg


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Description

Description

Description: The monoclonal antibody DE-U-10 recognizes desmin. Desmin is the protein subunit of class-III intermediate filaments. Desmin is found predominantly in skeletal, cardiac, and smooth muscle. Desmin plays a role in muscle cell development and differentiation, muscle cell architecture and structure, and mitochondrial function. Desmin forms scaffolds around the Z-disk of sarcomeres in muscle cells. Desmin knockout mice exhibit defects in skeletal, smooth, and cardiac muscle and have impaired mitochondrial function. Mutations in Desmin result in conditions such as desmin-related myopathy (DRM), cardiomyopathy dilated type 1I (CMD1I), and neurogenic scapuloperoneal syndrome Kaeser type (Kaeser syndrome).The DE-U-10 antibody recognizes human, mouse, rat, feline, bovine, goat, hamster, sheep, porcine, rabbit, and chicken desmin. The DE-U-10 antibody will also label tumors derived from muscle tissue, such as leiomyomas and rhabdomyosarcomas.Applications Reported: This DE-U-10 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, microscopy, and immunocytochemistry.Applications Tested: This DE-U-10 antibody has been tested by immunohistochemistry of formalin-fixed paraffin embedded human tissue using high pH antigen retrieval and can be used at less than or equal to 10 μg/mL. This DE-U-10 antibody has been tested by immunocytochemistry of fixed and permeabilized cells and can be used at less than or equal to 10 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome.Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Desmin
Monoclonal
0.2mg/mL
PBS with 0.09% sodium azide; pH 7.2
P48675, P02540, P31001, P17661, P02542, O62654
CMD1I, CSM2, DES, FLJ12025
Murine
Affinity chromatography
RUO
Antibody
Monoclonal
Sheep
Microscopy
DE-U-10
eFluor 660
DES
Conjugated
DES
IgG1, kappa
100μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
1674
Videos

For Research Use Only.