missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEPDC7, Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit Polyclonal Antibody
Brand: Abnova H00091614-D01.100uL
This item is not returnable.
View return policy
Description
Sequence: MATVQEKAAALNLSALHSPAHRPPGFSVAQKPFGATYVWSSIINTLQTQVEVKKRRHRLKRHNDCFVGSEAVDVIFSHLIQNKYFGDVDIPRAKVVRVCQALMDYKVFEAVPTKVFGKDKKPTFEDSSCSLYRFTTIPNQDSQLGKENKLYSPARYADALFKSSDIRSASLEDLWENLSLKPANSPHVNISATLSPQVINEVWQEETIGRLLQLVDLPLLDSLLKQQEAVPKIPQPKRQSTMVNSSNYLDRGILKAYSDSQEDEWLSAAIDCLEYLPDQMVVEISRSFPEQPDRTDLVKELLFDAIGRYYSSREPLLNHLSDVHNGIAELLVNGKTEIALEATQLLLKLLDFQNREEFRRLLYFMAVAANPSEFKLQKESDNRMVVKRIFSKAIVDNKNLSKGKTDLLVLFLMDHQKDVFKIPGTLHKIVSVKLMAIQNGRDPNRDAGYIYCQRIDQRDYSNNIEKTTKDELLNLLKTLDEDSKLSAKEKKKLLGQFYKCHPDIFIEHFGDSpecifications
DEPDC7 | |
Polyclonal | |
No additive | |
BC030970 | |
DEPDC7 | |
DEPDC7 (AAH30970.1, 1 a.a. ∼ 511 a.a) full-length human protein. | |
RUO | |
91614 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Immunoprecipitation, Western Blot | |
Unconjugated | |
DEP domain containing 7 | |
TR2/dJ85M6.4 | |
Rabbit | |
100 μL | |
Primary | |
Human | |
Antibody |