missing translation for 'onlineSavingsMsg'
Learn More

Paragon Scientific™ Density Standards (15 °C)

Paragon ISO 17025 / ISO 17034 certified Density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature within the range of 15°C

Brand:  Paragon Scientific™ DEN15-08

Additional Details : CAS Number : 64-17-5 Weight : 0.06000kg

 View more versions of this product

Product Code. 13460479

  • £56.45 / 60mL

Estimated Shipment
09-04-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Certified in accordance with primary level ASTM D1480 methodology
  • Ensures full compliance to ASTM and IP test method protocols.
  • Supplied in tamper evident packaging with a 2 year shelf life.
  • Fully traceable to international standards.
  • Supplied with MSDS.
  • Tamper evident security packaging.
  • Manufactured in the United Kingdom
Specifications

Specifications

Dual certified to ISO 17025 / ISO 17034 under our UKAS accreditation
0.9413 g/mL (15°C)
60mL
Videos
Documents

Nominal values quoted