Adam Equipment™ Density Determination Kit


Overview
Brand: Adam Equipment™

Manufacturer Part Number: 7452

Density kit Adam Equipment with thermometer &

UNSPSC: 41111516

Code: 03

Additional Details:
Additional Details: Weight: 1.00000kg



Product Code. 12763245

Quantity Price
1 £ 317.18 / Each
Estimated Shipment
24-02-2017
Add to basket


Description and Specification

Specification

Quantity 1 Each
Product Type Density Determination Kit
Includes Thermometer & holder, and beaker & holder replacement pan assembly
For Use With (Equipment) PW balances