Adam Equipment™ Density Determination Kit
Overview
Brand: Adam Equipment™
Manufacturer Part Number: 7453
UNSPSC: 41111516
Code: 03
Additional Details:
Additional Details: Weight: 1.00000kg
Product Code. 12753245
Quantity Price
1 £ 311.76 / Each
Estimated Shipment
12-06-2017
Add to basket
Description and Specification
Specification
Quantity 1 Each
Includes Thermometer & holder, and beaker & holder replacement pan assembly
For Use With (Equipment) PGW and PGL balances
Product Type Density Determination Kit
And beaker & holder replacement pan assembly included for PGW and PGL balances Adam