missing translation for 'onlineSavingsMsg'
Learn More

Dendritic Cell Marker DCIR2, PE, clone: 33D1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 12-5884-83

Code : Z2

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15218879

  • £162.21 / 200µg


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Description

Description

Description: The 33D1 monoclonal antibody reacts with a mouse dendritic cell (DC)-specific surface marker, DCIR2. The nature and biological activity of DCIR2 is unknown. DCIR2 has been reported on a variety of dendritic cell subpopulations from mouse thymus, spleen, lymph node, and Peyer's patch. Bone marrow dendritic cells require GM-CSF to express DCIR2, and this expression is downregulated in the presence of IL-4. DCIR2 has been detected in vivo in brain dendritic cells post infection with Toxoplasma gondii.Applications Reported: The 33D1 antibody has been reported for use in flow cytometric analysis.Applications Tested: The 33D1 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Dendritic Cell Marker DCIR2
Monoclonal
0.2mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Conjugated
Clec4a4
IgG2b, kappa
200μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary
474145
Flow Cytometry
33D1
PE
Clec4a4
DC Marker, DC inhibitor receptor 2, DCIR2
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
Videos

For Research Use Only.