missing translation for 'onlineSavingsMsg'
Learn More

Dendritic Cell Marker (33D1) Rat anti-Mouse, eFluor 660, Clone: 33D1, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 50-5884-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15559986

  • £60.53 / 25µg


Estimated Shipment date
from Supplier 04-04-2018
Add to basket

Description

Description

The 33D1 monoclonal antibody reacts with a mouse dendritic cell (DC)-specific surface marker, DCIR2. The nature and biological activity of DCIR2 is unknown. DCIR2 has been reported on a variety of dendritic cell subpopulations from mouse thymus, spleen, lymph node, and Peyer’s patch. Bone marrow dendritic cells require GM-CSF to express DCIR2, and this expression is downregulated in the presence of IL-4. DCIR2 has been detected in vivo in brain dendritic cells post infection with Toxoplasma gondii.
Specifications

Specifications

Dendritic Cell Marker (33D1)
33D1
eFluor 660
Conjugated
Rat
25μg
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
Flow Cytometry
0.2mg/mL
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
DC Marker, DC inhibitor receptor 2
IgG2b, kappa
RUO
Primary
Murine
Videos

For Research Use Only.