missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Denatonium benzoate, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
The bitterest compound knownDenatonium benzoate is used as an alcohol denaturant and flavor in pharmaceuticals. It is also used in antifreeze, nail biting preventions, respirator mask fit-testing, animal repellents, liquid soaps and shampoos. Further, it is used in air care products. It acts as H1 antihistamine. In addition to this, it is used as a disinfectant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
The bitterest compound knownDenatonium benzoate is used as an alcohol denaturant and flavor in pharmaceuticals. It is also used in antifreeze, nail biting preventions, respirator mask fit-testing, animal repellents, liquid soaps and shampoos. Further, it is used in air care products. It acts as H1 antihistamine. In addition to this, it is used as a disinfectant.
Solubility
Soluble in isopropyl alcohol, methanol. Slightly soluble in acetone. Insoluble in ether.
Notes
Incompatible with strong oxidizing agents. Moisture sensitive.
The bitterest compound knownDenatonium benzoate is used as an alcohol denaturant and flavor in pharmaceuticals. It is also used in antifreeze, nail biting preventions, respirator mask fit-testing, animal repellents, liquid soaps and shampoos. Further, it is used in air care products. It acts as H1 antihistamine. In addition to this, it is used as a disinfectant.
Solubility
Soluble in isopropyl alcohol, methanol. Slightly soluble in acetone. Insoluble in ether.
Notes
Incompatible with strong oxidizing agents. Moisture sensitive.
Specifications
Quantity | 1 g |
Solubility | Soluble in isopropyl alcohol,methanol. Slightly soluble in acetone. Insoluble in ether. |
Merck Index | 142891 |
Melting Point | 164°C to 169°C |
Formula Weight | 446.59 |
Physical Form | Solid |
Color | White |
Chemical Name or Material | Denatonium benzoate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'