missing translation for 'onlineSavingsMsg'
Learn More

Delavirdine mesylate, Tocris Bioscience™

Non-nucleoside reverse transcriptase inhibitor

Brand:  RD Systems (Bio-Techne) 4149/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13454244

  • £457.17 / 50mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Non-nucleoside reverse transcriptase inhibitor (NNRTI). Selectively inhibits HIV-1 reverse transcriptase (RNA-dependent DNA polymerase) over other cellular polymerases (IC50 values are 0.26, 440 and >550 μM for HIV-1 reverse transcriptase, DNA polymerase α and DNA polymerase δ). Prevents HIV-1 spread in vivo and blocks viral replication in peripheral blood lymphocytes. Orally active.
Specifications

Chemical Identifiers

147221-93-0
552.665
delavirdine mesylate, delavirdine mesilate, rescriptor, unii-421105krqe, delavirdine monomethanesulfonate, delavirdine mesylate usan, rescriptor tn, 1-3-isopropylamino-2-pyridyl-4-5-methanesulfonamidoindol-2-yl carbonyl piperazine monomethanesulfonate
CHEBI:4379
CC(C)NC1=C(N=CC=C1)N2CCN(CC2)C(=O)C3=CC4=C(N3)C=CC(=C4)NS(=O)(=O)C.CS(=O)(=O)O
C23H32N6O6S2
MEPNHSOMXMALDZ-UHFFFAOYSA-N
441386
methanesulfonic acid;N-[2-[4-[3-(propan-2-ylamino)pyridin-2-yl]piperazine-1-carbonyl]-1H-indol-5-yl]methanesulfonamide

Specifications

50mg
147221-93-0
delavirdine mesylate, delavirdine mesilate, rescriptor, unii-421105krqe, delavirdine monomethanesulfonate, delavirdine mesylate usan, rescriptor tn, 1-3-isopropylamino-2-pyridyl-4-5-methanesulfonamidoindol-2-yl carbonyl piperazine monomethanesulfonate
CC(C)NC1=C(N=CC=C1)N2CCN(CC2)C(=O)C3=CC4=C(N3)C=CC(=C4)NS(=O)(=O)C.CS(=O)(=O)O
552.665
CHEBI:4379
>99%
Delavirdine mesylate
C23H32N6O6S2
MEPNHSOMXMALDZ-UHFFFAOYSA-N
methanesulfonic acid;N-[2-[4-[3-(propan-2-ylamino)pyridin-2-yl]piperazine-1-carbonyl]-1H-indol-5-yl]methanesulfonamide
441386
552.67
Videos
Documents