Learn More
Invitrogen™ Dehydroepiandrosterone Sulfate (DHEA-S) Competitive ELISA Kit
Competitive ELISA
Brand: Invitrogen™ EIADHEA
Code : NEW
Additional Details : Weight : 0.33820kg
Includes: Pre-coated plate(s), standard, detector antibody, HRP conjugate, assay buffer, wash buffer, chromogen, stop solution, plate covers and lot-specific technical data sheet.
Description
The Dehydroepiandrosterone Sulfate (DHEA-S) ELISA quantitates DHEA-Se in serum,plasma,urine,saliva,dried fecal extracts or cell culture medium.
Principle of the method
The Dehydroepiandrosterone sulfate (DHEA-S) Competitive ELISA research-use-only kit uses a specifically generated antibody to measure Dehydroepiandrosterone sulfate (DHEA-S) in serum,plasma,urine,and saliva samples,and in fecal extracts. The kit will also quantitatively measure DHEA-S present in tissue culture media samples. A DHEA-S standard is provided to generate a standard curve for the assay and all samples should be read off the standard curve. Standards or diluted samples are pipetted into a clear microtiter plate coated with an antibody to capture sheep antibodies. A DHEA-S-peroxidase conjugate is added to the standards and samples in the wells. The binding reaction is initiated by the addition of a polyclonal antibody to DHEA-S to each well. After a 2-hour incubation the plate is washed and substrate is added. The substrate reacts with the bound DHEA-S-peroxidase conjugate. After a short incubation,the reaction is stopped and the intensity of the generated color is detected in a microtiter plate reader capable of measuring at 450 nm.
Rigorous validation
Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity,specificity,precision,and lot-to-lot consistency. See manual for more information on validation.
Specifications
96-60,000 pg/mL | |
Dehydroepiandrosterone sulfate (DHEA-S) Competitive ELISA Kit | |
HRP | |
ELISA Kit | |
Fecal Extract, Plasma, Saliva, Serum, Supernatant, Urine | |
Human | |
Colorimetric Microplate Reader | |
7% | |
HRP | |
RUO | |
Fecal Extract,50 μL; Plasma, 1 μL; Saliva,25 μL; Serum, 1 μL; Supernatant, 50 μL; Urine,25 μL | |
Chemical | |
2 hr. 30 min. |
90.9 pg/mL | |
Multiplex assay on the Luminex Platform | |
Fluorescence | |
Steroid hormone | |
96 assays | |
ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti | |
8.4% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
96 Tests | |
Reproductive Biology | |
-20°C | |
1 hr. 20 min. |