missing translation for 'onlineSavingsMsg'
Learn More

Dehydroacetic acid sodium salt, 99%, Thermo Scientific Chemicals

Catalog Number p-2691305
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
4418-26-2
C8H7O4Na
MFCD00040583
3-Acetyl-6-methyl-2H-pyran-2, 4(3H)-dione sodium salt
This item is not returnable. View return policy
4418-26-2
C8H7O4Na
MFCD00040583
3-Acetyl-6-methyl-2H-pyran-2, 4(3H)-dione sodium salt

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 295.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Quantity 500 g
Formula Weight 190.13
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Dehydroacetic acid sodium salt, 99%
compliance-icons
Product Identifier
  • Dehydroacetic acid sodium salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'