Learn More
OHAUS™ Defender™ 5000 R Series Base
OHAUS™ Defender™ 5000 R Series Base offers durability for demanding industrial applications. The R series features moisture-protected single-point load cells, powder-coated steel construction, non-slip rubber adjustable feet, removable stainless steel weighing pans and leveling bubbles.
Brand:  OHAUS™ 30419618
Additional Details : Weight : 31.00000kg
 View more versions of this product
Product Code. 15948425
  • £1065.00 / 1 unit
Estimated Shipment: 07-05-2024
to see stock.
Description
Description
  • The R Series features a stainless steel platform, powder-coated steel frame and an aluminum load cell to withstand harsh industrial environments.
  • The Defender 5000 bases are equipped with an EasyConnect cable, which allows instant configuration with Defender 5000 (TD52) indicators, speeding up installation.
  • Defender 5000 bases are NTEP-certified and Measurement Canada approved.
  • These bases also carry OIML and EC Type approvals, which make them ideal for legal-for-trade use.
Specifications
Specifications
304-grade stainless steel platform, sand grey powder-coated steel frame, aluminum load cell, non-slip adjustable rubber feet, level indicator
10.000000 g, 20.000000 g
IP67
633 x 610 x 130 mm
150.00 kg, 300.00 kg
D300RQDV
-10°C to +40°C
31.000 kg