Learn More
OHAUS™ Defender™ 5000 - D52 Bench Scale, Models TD52XW RTV
OHAUS™ Defender™ 5000 Series multifunctional bench scale is ideal for a multitude of applications. Built durably and equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management and multiple connectivity options.
Brand:  OHAUS™ 30461589
Additional Details : Weight : 48.00000kg
 View more versions of this product
Product Code. 15948595
  • £1955.00 / 1 unit
Estimated Shipment: 16-04-2024
to see stock.
Description
Description
  • User-friendly model with an easy-to-use keypad, large backlit display and plain text on-screen prompts which guide you through operation.
  • Durable construction with a stainless steel platform, sand gray powder-coated steel frame and aluminum load cell.
  • Defender meets NTEP/Measurement Canada or OIML/EC type approvals (depending on region).
  • Features common industrial modes and supports a 50000-item library with 1000 user profiles for easy data management.
  • Includes GMP/GLP with time/date for result traceability.
Specifications
Specifications
20 Hours with Rechargeable Battery
Stainless steel (TD52XW) indicator, 304-grade stainless steel platform with sand gray powder-coated steel frame, aluminum load cell, non-slip adjustable rubber feet
Backlit LCD, 256 x 80 pixels
D52XW150RTDV3-M
Gram, Kilogram, Metric Tonnes
48.000 kg
Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock
60.00 kg, 150.00 kg
20.000000 g, 50.000000 g
-10°C to +40°C
963 x 600 x 1234 mm