Learn More
Ohaus™ Defender™ 3000 Bench Scale
Ideal for general weighing and simple counting applications when working in a damp environment or when it is essential to thoroughly clean the scale.
Brand: Ohaus™ 80250493
Code : A0
Additional Details : Weight : 24.50000kg Transport : UN number : 2800 Chem class : 8
Description
- Simple, yet rugged tubular frame design.
- Indicators with tactile keys.
- Backlit LCD display.
- Built-in RS232 bidirectional interface.
- Internal rechargeable sealed lead-acid battery.
- Economy line of dry or wet-use indicators, bases and bench scales is ideal for general weighing and simple counting applications in production, packaging, warehouse, inventory, shipping and receiving areas.
Specifications
100 Hours with Rechargeable Battery | |
ABS indicator with reversible rear cover, painted steel column and wall bracket, 304 grade stainless steel platform with painted steel tubular frame, IP67 aluminum load cell, self-adjusting stainless steel feet with removable rubber caps | |
LCD with White Backlight | |
RS232, AC Adapter, Rechargeable Battery | |
D31P60BL-M | |
668.00 x 420.00 x 920.000 mm | |
668.00 mm | |
16.50 in. | |
420 mm | |
920.000 mm | |
668.00 mm |
RS232 (Included) | |
60 kg | |
20.000000 g | |
ABS indicator with reversible rear cover, painted steel column and wall bracket, 304 grade stainless steel platform with painted steel tubular frame, IP67 aluminum load cell, self-adjusting stainless steel feet with removable rubber caps | |
668.00 x 420.00 mm | |
26.30 in. | |
550 mm | |
420.00 mm | |
36.20 in. | |
26.30 in. |