missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Dectin-1/CLEC7A Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
POA
Specifications
Antigen | Dectin-1/CLEC7A |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready |
Applications | Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF |
Classification | Polyclonal |
Description
Dectin-1/CLEC7A Polyclonal specifically detects Dectin-1/CLEC7A in Mouse samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Specifications
Dectin-1/CLEC7A | |
Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
Polyclonal | |
Goat | |
Mouse | |
Beta-glucan receptor, BGR, CD369, CLEC7A, CLECSF12, CLECSF12DC-associated C-type lectin 1, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 12, C-type lectin domain family 7 member A, C-type lectin domain family 7, member A, C-type lectin superfamily member 12, dectin-1, Dectin1, DECTIN1CANDF4, Dendritic cell-associated C-type lectin 1, dendritic cell-associated C-type lectin-1, hDectin-1, lectin-like receptor 1 | |
CLEC7A | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
Unconjugated | |
RUO | |
Q6QLQ4 | |
64581 | |
Mouse myeloma cell line NS0-derived recombinant mouse Dectin-1/CLEC7A Phe69-Leu244 Accession # Q6QLQ4 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Dectin-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human Dectin-1 and recombinant mouse Dectin-2 alpha is observed. |
For Research Use Only