missing translation for 'onlineSavingsMsg'
Learn More

Decon Labs™ Decomatic

Brand:  Decon Labs™ DEC5

Code : 04

Additional Details : Weight : 5.20000kg


Quantity 5000mL

 View more versions of this product

Product Code. 12327143

  • £48.40 / 5 L


In Stock 46
Add to basket


Description

Description

Biodegradable, phosphate, enzyme and chlorine free, suitable for automatic washing machines. By preventing the deposition of lime and salts on both contents and machine parts, it minimises corrosion and wear of seals, pumps, racks and stainless steel surfaces, thereby providing more efficient performance and longer life to the machine. Not suitable for use on non-ferrous metals, notably aluminium and zinc, or on polycarbonate. Ideal for use on glassware and equipment in medical, pharmaceutical, research, educational, scientific and industrial laboratories.
Specifications

Chemical Identifiers

Yes
Yes
Pale amber
>13
No
2.28cps at 20°C
Liquid

Specifications

Yes
No
2.28cps at 20°C
>13
Liquid
Yes
Pale amber
5000mL
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • DECON DECOMATIC

Signal Word
  • Warning

Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H290-May be corrosive to metals.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Supplemental information
  • MIXTURE LIST-Contains¬†: NaOH

SDS Documentation