Learn More
Corning™ DeckWorks™ Low-Binding Hinged Rack Pipet Tips, Sterile
Designed for quantitative assays in which sample binding can significantly alter results
£80.97 - £419.21
Specifications
Sterility | Sterile |
---|---|
Color | Natural |
For Use With (Application) | Designed for quantitative assays in which sample binding can significantly alter results |
Format | Hinged Rack |
Tip Style | Low Retention, Round |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11353675
|
Corning™
4151 |
1 to 200 μL |
£419.21
Pack of 3840 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||||
11719335
|
Corning™
4152 |
1 to 300 μL |
£104.80
Pack of 960 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||||
11799325
|
Corning™
4150 |
0.1 to 10 μL |
£104.80
Pack of 960 |
Estimated Shipment: 10-09-2024
Log in to see stock. |
|||||
11363675
|
Corning™
4152 |
1 to 300 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10291440
|
Corning™
4150 |
0.1 to 10 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11709335
|
Corning™
4151 |
1 to 200 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Corning DeckWorks low binding tips are available in 10 μL, 200 μL, and 300 μL sizes. Proprietary low binding technology reduces surface tension on the interior wall of the tip. Leaching and/or subsequent sample degradation associated with silicone-based tips are eliminated. DeckWorks low binding tips are ideal for quantitative assays in which sample binding can significantly alter results. Specific applications include accurate and precise pipetting of expensive reagents, DNA, proteins and/or peptides.
Specifications
Sterile | |
Designed for quantitative assays in which sample binding can significantly alter results | |
Low Retention, Round | |
Yes | |
Polypropylene |
Natural | |
Hinged Rack | |
Hinged rack | |
DeckWorks |