missing translation for 'onlineSavingsMsg'
Learn More

Decanoic acid, 99%, Thermo Scientific Chemicals

Catalog Number 10722211
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Glass bottle
Plastic Drum
This item is not returnable. View return policy
334-48-5
C10H20O2
172.268
MFCD00004441
GHVNFZFCNZKVNT-UHFFFAOYSA-N
capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid
2969
CHEBI:30813
decanoic acid
CCCCCCCCCC(=O)O
This item is not returnable. View return policy
334-48-5
C10H20O2
172.268
MFCD00004441
GHVNFZFCNZKVNT-UHFFFAOYSA-N
capric acid, n-decanoic acid, n-capric acid, decylic acid, caprinic acid, decoic acid, n-decylic acid, n-decoic acid, 1-nonanecarboxylic acid, caprynic acid
2969
CHEBI:30813
decanoic acid
CCCCCCCCCC(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 30.0°C to 32.0°C
Boiling Point 268.0°C to 270.0°C
Color White
Assay Percent Range 99%
Linear Formula CH3(CH2)8CO2H
Beilstein 02, II, 309
Merck Index 15, 1760
Solubility Information Solubility in water: 0.15g/L (20°C). Other solubilities: soluble in dilute hno3, soluble in ethanol, ether, chloroform, benzene and, carbon disulfide
Formula Weight 172.27
Percent Purity 99%
Density 0.8900g/mL
Quantity 500 g
Flash Point 150°C
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Crystalline Solid
Specific Gravity 0.89
Chemical Name or Material Decanoic acid, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'