Deb USA Universal Protect Pre-work Skin Cream


Overview
Brand: Deb USA

Manufacturer Part Number: UPW150ML

UNSPSC: 53131613

Code: 04

Additional Details:
Additional Details: Weight: 1.00000kg



Product Code. 12829247

Quantity Price
1 £ 50.85 / Pack of 12
Estimated Shipment
21-04-2017
Add to basket


Description and Specification

Specification

Form Cream
Product Type Universal Protect Skin Cream
Color Pale yellow
Shelf Life 24 Month(s)
Size (Metric) 147mL
Packaging Tube
Quantity 12 Case
pH Range 6 to7.5
Composition Water, Kaolin, Cetearyl Alcohol, Dicaprylyl Carbonate, Glycerin, Xanthan Gum, Phenoxyethanol, Sodium Cetearyl Sulfate, Sodium Lauryl Sulfate, Methylparaben, 2-Bromo-2-Nitropropane-1,3-diol, Butylparaben, Ethylparaben, Isobutylparaben, Propylparaben, Yellow 5, Yellow 6
For Use With (Application) Helps protect the skin against contact with various workplace contaminants and working environments such as wet or dry condition